trans-4-(Boc-amino)cyclohexanol ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

B3564-1G B3564-5G
TCB3564-1GEA 44.24 CAD
TCB3564-1G TCB3564-5G
trans-4-(Boc-amino)cyclohexanol ≥98.0% (by GC, total nitrogen)
trans-4-(Boc-amino)cyclohexanol

CAS Number: 111300-06-2
Molecular Formula: C11H21NO3
Molecular Weight: 215.29
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 171

Formula: C₁₁H₂₁NO₃
Melting Pt: 171 °C
MDL Number: MFCD03844613
CAS Number: 111300-06-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d4c0d27a812f17846d972fa3cda02a3639c1430&vid=5d4c0d27a812f17846d972fa3cda02a3639c1430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR