9-Benzylcarbazole ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
B3558-25G
B3558-5G
TCB3558-25GEA
558.69
CAD
TCB3558-25G
TCB3558-5G
9-Benzylcarbazole ≥98.0% (by GC, total nitrogen)
9-Benzylcarbazole
CAS Number: 19402-87-0
Molecular Formula: C19H15N
Molecular Weight: 257.34
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 268
Melting point (°C): 119
Lambda max.: 341 nm (MeOH)
Formula:
C₁₉H₁₅N Boiling Pt: 268 °C Melting Pt: 119 °C |
MDL Number:
MFCD00218286 CAS Number: 19402-87-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...