9-(4-Bromophenyl)carbazole ≥98.0%

Supplier: TCI America

Synonyms: 9-(4-Bromophenyl)-9H-carbazole

B3554-1G B3554-5G
TCB3554-1GEA 66.36 CAD
TCB3554-1G TCB3554-5G
9-(4-Bromophenyl)carbazole ≥98.0%
9-(4-Bromophenyl)carbazole

CAS Number: 57102-42-8
MDL Number: MFCD11617969
Molecular Formula: C18H12BrN
Molecular Weight: 322.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 149

Formula: C₁₈H₁₂BrN
Melting Pt: 149 °C
MDL Number: MFCD11617969
CAS Number: 57102-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=505d77f0f00141213af32143b062d1b4c4e878d4&vid=505d77f0f00141213af32143b062d1b4c4e878d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR