2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,7-Dibromo-9,9-bis(6-bromohexyl)-9H-fluorene

B3552-1G B3552-5G
TCB3552-1GEA 388.68 CAD
TCB3552-1G TCB3552-5G
2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene ≥98.0% (by HPLC)
2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene

CAS Number: 570414-33-4
MDL Number: MFCD09260450
Molecular Formula: C25H30Br4
Molecular Weight: 650.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 72

Formula: C₂₅H₃₀Br₄
MW: 650.13 g/mol
Melting Pt: 72 °C
MDL Number: MFCD09260450
CAS Number: 570414-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=769b54e5eb1843e301533fabcff97d519cf2d672&vid=769b54e5eb1843e301533fabcff97d519cf2d672&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR