1,3-Bis(3-aminophenoxy)-5-chlorobenzene dihydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

B3550-1G B3550-5G
TCB3550-1GEA 347.6 CAD
TCB3550-1G TCB3550-5G
1,3-Bis(3-aminophenoxy)-5-chlorobenzene dihydrochloride ≥98.0% (by HPLC)
1,3-Bis(3-aminophenoxy)-5-chlorobenzene dihydrochloride

CAS Number: 99503-73-8
Molecular Formula: C18H15ClN2O2
Molecular Weight: 399.70
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₁₈H₁₅ClN₂O₂
MDL Number: MFCD14708170
CAS Number: 99503-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64ce8612d59aaf3267514ddf5883cf93d6ae8cce&vid=64ce8612d59aaf3267514ddf5883cf93d6ae8cce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR