1-Bromo-3,5-di-tert-butylbenzene ≥98.0% (by GC)
Supplier: TCI America
B3547-25G
B3547-5G
TCB3547-25GEA
472.74
CAD
TCB3547-25G
TCB3547-5G
1-Bromo-3,5-di-tert-butylbenzene ≥98.0% (by GC)
1-Bromo-3,5-di-tert-butylbenzene
CAS Number: 22385-77-9
MDL Number: MFCD00796945
Molecular Formula: C14H21Br
Molecular Weight: 269.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 65
Formula:
C₁₄H₂₁Br Boiling Pt: 158 °C Melting Pt: 65 °C |
MDL Number:
MFCD00796945 CAS Number: 22385-77-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...