Bis(tricyclohexylphosphine)nickel(II) dichloride ≥95.0%

Supplier: TCI America

Synonyms: Bis(tricyclohexylphosphine)nickel(II) chloride, Dichlorobis(tricyclohexylphosphine)nickel(II)

B3534-1G B3534-5G
TCB3534-1GEA 62.68 CAD
TCB3534-1G TCB3534-5G
Bis(tricyclohexylphosphine)nickel(II) dichloride ≥95.0%
Bis(tricyclohexylphosphine)nickel(II) dichloride

CAS Number: 19999-87-2
MDL Number: MFCD11973802
Molecular Formula: C36H66Cl2NiP2
Molecular Weight: 690.46
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 227


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Dichlorobis(tricyclohexylphosphine)nickel(II)

Formula: C₃₆H₆₆Cl₂NiP₂
MW: 690.46 g/mol
Melting Pt: 227 °C
MDL Number: MFCD11973802
CAS Number: 19999-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be1873ee5433f51e9704e92bdd28cdeb26504da4&vid=be1873ee5433f51e9704e92bdd28cdeb26504da4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR