2,2'-Bipyridine-6,6'-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3533-1G
B3533-5G
TCB3533-1GEA
369.72
CAD
TCB3533-1G
TCB3533-5G
2,2'-Bipyridine-6,6'-dicarboxylic acid ≥98.0% (by GC, titration analysis)
2,2'-Bipyridine-6,6'-dicarboxylic acid
CAS Number: 4479-74-7
MDL Number: MFCD11042476
Molecular Formula: C12H8N2O4
Molecular Weight: 244.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 288
Lambda max.: 288 nm (H2O)
Formula:
C₁₂H₈N₂O₄ Melting Pt: 288 °C |
MDL Number:
MFCD11042476 CAS Number: 4479-74-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...