3-Bromo-5-methylanisole ≥98.0% (by GC)

Supplier: TCI America

B3526-1G B3526-5G
TCB3526-1GEA 36.34 CAD
TCB3526-1G TCB3526-5G
3-Bromo-5-methylanisole ≥98.0% (by GC)
3-Bromo-5-methylanisole

CAS Number: 29578-83-4
MDL Number: MFCD08061916
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 1.42


Other Product Names / Synonyms: 3-Bromo-5-methylanisole

Formula: C₈H₉BrO
Boiling Pt: 122 °C
Density: 1.42 (20 °C)
MDL Number: MFCD08061916
CAS Number: 29578-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c04e0e93f0d129da695543e7f97c9e31043247f0&vid=c04e0e93f0d129da695543e7f97c9e31043247f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR