2-Bromodibenzothiophene ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromodibenzo[b,d]thiophene

B3525-25G B3525-5G
TCB3525-25GEA 451.25 CAD
TCB3525-25G TCB3525-5G
2-Bromodibenzothiophene ≥95.0% (by GC)
2-Bromodibenzothiophene

CAS Number: 22439-61-8
MDL Number: MFCD00089285
Molecular Formula: C12H7BrS
Molecular Weight: 263.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 125

Formula: C₁₂H₇BrS
MW: 263.15 g/mol
Melting Pt: 125 °C
MDL Number: MFCD00089285
CAS Number: 22439-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c0584535da5ce95d5fd626a9bdd46a49d5915ee&vid=1c0584535da5ce95d5fd626a9bdd46a49d5915ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR