2-Bromo-4-nitropyridine ≥95.0% (by GC)

Supplier: TCI America

B3523-1G B3523-5G
TCB3523-1GEA 53.72 CAD
TCB3523-1G TCB3523-5G
2-Bromo-4-nitropyridine ≥95.0% (by GC)
2-Bromo-4-nitropyridine

CAS Number: 6945-67-1
MDL Number: MFCD03701383
Molecular Formula: C5H3BrN2O2
Molecular Weight: 203.00
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 62

Formula: C₅H₃BrN₂O₂
Melting Pt: 62 °C
MDL Number: MFCD03701383
CAS Number: 6945-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae6da7af643f6f9193e28ba7060794f3e7612bcc&vid=ae6da7af643f6f9193e28ba7060794f3e7612bcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR