4-Bromo-3-methyl-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI America
B3522-1G
B3522-5G
TCB3522-1GEA
33.18
CAD
TCB3522-1G
TCB3522-5G
4-Bromo-3-methyl-1H-pyrazole ≥98.0% (by GC)
4-Bromo-3-methyl-1H-pyrazole
CAS Number: 13808-64-5
MDL Number: MFCD00005241
Molecular Formula: C4H5BrN2
Molecular Weight: 161.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76
Formula:
C₄H₅BrN₂ Melting Pt: 76 °C |
MDL Number:
MFCD00005241 CAS Number: 13808-64-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...