4-Bromo-3,5-dimethylisoxazole ≥98.0% (by GC)

Supplier: TCI America

B3519-1G B3519-5G
TCB3519-1GEA 33.18 CAD
TCB3519-1G TCB3519-5G
4-Bromo-3,5-dimethylisoxazole ≥98.0% (by GC)
4-Bromo-3,5-dimethylisoxazole

CAS Number: 10558-25-5
MDL Number: MFCD00068187
Molecular Formula: C5H6BrNO
Molecular Weight: 176.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Specific Gravity (20/20): 1.54
Storage Temperature: 0-10°C

Formula: C₅H₆BrNO
MW: 176.01 g/mol
Boiling Pt: 75 °C
Density: 1.54 (20 °C)
MDL Number: MFCD00068187
CAS Number: 10558-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96b286cec4d3faf439f676ad4e939b2c1e73e349&vid=96b286cec4d3faf439f676ad4e939b2c1e73e349&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR