2-Bromo-1,3,5-triisopropylbenzene ≥95.0% (by GC)

Supplier: TCI America

B3513-25G B3513-5G
TCB3513-25GEA 118.97 CAD
TCB3513-25G TCB3513-5G
2-Bromo-1,3,5-triisopropylbenzene ≥95.0% (by GC)
2-Bromo-1,3,5-triisopropylbenzene

CAS Number: 21524-34-5
MDL Number: MFCD00051547
Molecular Formula: C15H23Br
Molecular Weight: 283.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 110
Specific Gravity (20/20): 1.13

Formula: C₁₅H₂₃Br
Boiling Pt: 148 °C
Density: 1.13 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00051547
CAS Number: 21524-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8837ec13213d14e4b20563d6bb747da1bee3118&vid=c8837ec13213d14e4b20563d6bb747da1bee3118&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR