4,4'-Bis(hydroxymethyl)-2,2'-bipyridine ≥95.0%

Supplier: TCI America

Synonyms: 4,4'-Bis(hydroxymethyl)-2,2'-bipyridyl, [2,2'-Bipyridine]-4,4'-diyldimethanol, 2,2'-Bipyridine-4,4'-dimethanol

B3511-1G B3511-5G
TCB3511-1GEA 305.73 CAD
TCB3511-1G TCB3511-5G
4,4'-Bis(hydroxymethyl)-2,2'-bipyridine ≥95.0%
4,4'-Bis(hydroxymethyl)-2,2'-bipyridine

CAS Number: 109073-77-0
MDL Number: MFCD06202808
Molecular Formula: C12H12N2O2
Molecular Weight: 216.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 174


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 2,2'-Bipyridine-4,4'-dimethanol, 4,4'-Bis(hydroxymethyl)-2,2'-bipyridyl

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
Melting Pt: 174 °C
MDL Number: MFCD06202808
CAS Number: 109073-77-0

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR