2,2'-Bicinchoninic acid ≥98.0%
Supplier: TCI America
Synonyms:
[2,2'-Biquinoline]-4,4'-dicarboxylic acid, 2,2'-Biquinoline-4,4'-dicarboxylic Acid, 2,2′-Bicinchoninic acid
CAS Number: 1245-13-2
MDL Number: MFCD00068342
Molecular Formula: C20H12N2O4
Molecular Weight: 344.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 365
Product Categories: Materials Chemistry, Solar Cell Research Reagents, Ligands for Functional Metal Complexes, Bioscience, Protein Quantitative Assay Reagents, Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 2,2'-Biquinoline-4,4'-dicarboxylic Acid
Formula:
C₂₀H₁₂N₂O₄ MW: 344.33 g/mol Melting Pt: 365 °C |
MDL Number:
MFCD00068342 CAS Number: 1245-13-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...