2,2'-Bipyridine-5,5'-dicarboxylic acid 98,0
Supplier: TCI America
|
Warning
|
Synonyms:
6,6′-Dinicotinic acid
B3502-1G
B3502-5G
TCB3502-1GEA
191.18
CAD
TCB3502-1G
TCB3502-5G
2,2'-Bipyridine-5,5'-dicarboxylic acid 98,0
2,2'-Bipyridine-5,5'-dicarboxylic acid
CAS Number: 1802-30-8
MDL Number: MFCD01318320
Molecular Formula: C12H8N2O4
Molecular Weight: 244.21
Purity/Analysis Method:
Form: Crystal
Formula:
C12H8N2O4 MW: 244.21 g/mol Melting Pt: 365 °C |
MDL Number:
MFCD01318320 CAS Number: 1802-30-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...