(5-Bromo-3-pyridinyl)methanol ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromo-5-(hydroxymethyl)pyridine, 5-Bromo-3-pyridinemethanol
B3493-1G
B3493-5G
TCB3493-1GEA
148.52
CAD
TCB3493-1G
TCB3493-5G
(5-Bromo-3-pyridinyl)methanol ≥98.0%
(5-Bromo-3-pyridinyl)methanol
CAS Number: 37669-64-0
MDL Number: MFCD03265757
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.67
Other Product Names / Synonyms: 3-Bromo-5-(hydroxymethyl)pyridine
Formula:
C₆H₆BrNO Density: 1.67 (20 °C) |
MDL Number:
MFCD03265757 CAS Number: 37669-64-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...