3-Bromo-5-formylpyridine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde
B3490-1G
B3490-5G
TCB3490-1GEA
54.51
CAD
TCB3490-1G
TCB3490-5G
3-Bromo-5-formylpyridine ≥98.0% (by GC)
3-Bromo-5-formylpyridine
CAS Number: 113118-81-3
MDL Number: MFCD03265758
Molecular Formula: C6H4BrNO
Molecular Weight: 186.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98
Other Product Names / Synonyms: 3-Bromo-5-formylpyridine, 5-Bromonicotinaldehyde
Formula:
C₆H₄BrNO MW: 186.01 g/mol Melting Pt: 98 °C |
MDL Number:
MFCD03265758 CAS Number: 113118-81-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...