(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, tert-Butyl ((1S,2R)-2-aminocyclohexyl)carbamate, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
CAS Number: 365996-30-1
MDL Number: MFCD09952105
Molecular Formula: C11H22N2O2
Molecular Weight: 214.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Specific rotation [a]20/D: -6.0 deg (C=1, CH2Cl2)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol |
MDL Number:
MFCD09952105 CAS Number: 365996-30-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...