Bromoferrocene ≥95.0% (by GC, titration analysis)

Supplier: TCI America

B3476-1G B3476-5G
TCB3476-1GEA 246.48 CAD
TCB3476-1G TCB3476-5G
Bromoferrocene ≥95.0% (by GC, titration analysis)
Bromoferrocene

CAS Number: 1273-73-0
MDL Number: MFCD03093913
Molecular Formula: C10H9BrFe
Molecular Weight: 264.93
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 34

Formula: C₁₀H₉BrFe
Melting Pt: 34 °C
MDL Number: MFCD03093913
CAS Number: 1273-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2733e373b00f35cfe361b698e66fb7c5c2078d53&vid=2733e373b00f35cfe361b698e66fb7c5c2078d53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR