5-Bromo-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research

Supplier: TCI America

Synonyms: Bluo-Gal, (2S,3R,4S,5R,6R)-2-((5-Bromo-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 5-Bromo-3-indolyl β-D-galactopyranoside

B3470-100MG B3470-20MG
TCB3470-100MGEA 72.35 CAD
TCB3470-100MG TCB3470-20MG
5-Bromo-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research
5-Bromo-3-indolyl-beta-D-galactopyranoside

[for Biochemical Research]
CAS Number: 97753-82-7
MDL Number: MFCD00063691
Molecular Formula: C14H16BrNO6
Molecular Weight: 374.19
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -34.5 deg (C=1, DMF:H2O=1:1)
Storage Temperature: <0°C


Product Categories: Bioscience, Galactose, ƒÀ-Galactosidase Substrates
Other Product Names / Synonyms: Bluo-Gal

Formula: C₁₄H₁₆BrNO₆
MW: 374.19 g/mol
MDL Number: MFCD00063691
CAS Number: 97753-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb057b15a790e33e4a95a8b1aeaadd87254b9de8&vid=f022c900162d7e5be00c0a85a208dfa89bd19dc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR