2-(3-Bromopropyl)-1,3-dioxolane ≥97.0% (by GC)

Supplier: TCI America

B3463-5G B3463-1G
TCB3463-5GEA 587.76 CAD
TCB3463-5G 76219-366
2-(3-Bromopropyl)-1,3-dioxolane ≥97.0% (by GC)
2-(3-Bromopropyl)-1,3-dioxolane

CAS Number: 62563-07-9
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 1.44
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Bromobutyraldehyde Ethylene Acetal

Formula: C₆H₁₁BrO₂
Boiling Pt: 88 °C
Density: 1.44 (20 °C)
MDL Number: MFCD09868694
CAS Number: 62563-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5047ff5f32eefdd59de3ea05856d96ff0616ce77&vid=5047ff5f32eefdd59de3ea05856d96ff0616ce77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR