3-Benzylamino-1-propanol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-(Benzylamino)propan-1-ol, N-(3-Hydroxypropyl)benzylamine, 3-(Benzylamino)-1-propanol

B3462-5G B3462-25G
TCB3462-5GEA 74.26 CAD
TCB3462-5G TCB3462-25G
3-Benzylamino-1-propanol ≥98.0% (by titrimetric analysis)
3-Benzylamino-1-propanol

CAS Number: 4720-29-0
MDL Number: MFCD00020622
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: N-(3-Hydroxypropyl)benzylamine

Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 115 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00020622
CAS Number: 4720-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e94c5589df8ade930bc9651903d716868057d3c6&vid=e94c5589df8ade930bc9651903d716868057d3c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR