3-Bromocarbazole ≥97.0% (by GC)

Supplier: TCI America

B3458-5G B3458-25G
TCB3458-5GEA 97.49 CAD
TCB3458-5G TCB3458-25G
3-Bromocarbazole ≥97.0% (by GC)
3-Bromocarbazole

CAS Number: 1592-95-6
MDL Number: MFCD00222621
Molecular Formula: C12H8BrN
Molecular Weight: 246.11
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 198

Formula: C₁₂H₈BrN
Boiling Pt: 160 °C
Melting Pt: 198 °C
MDL Number: MFCD00222621
CAS Number: 1592-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bea2389f5e1642d4b32acf1476a12f4d1f1ad6b&vid=3bea2389f5e1642d4b32acf1476a12f4d1f1ad6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR