1-Boc-3-piperidone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-BOC-3-piperidone, 1-Boc-3-Piperidinone

B3309-25G B3309-5G
TCB3309-25GEA 192.76 CAD
TCB3309-25G TCB3309-5G
1-Boc-3-piperidone ≥98.0% (by GC)
1-Boc-3-piperidone

CAS Number: 98977-36-7
MDL Number: MFCD01631193
Molecular Formula: C10H17NO3
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 38


Other Product Names / Synonyms: 1-Boc-3-piperidone, tert-Butyl 3-Oxo-1-piperidinecarboxylate, 3-Oxo-1-piperidinecarboxylic Acid tert-Butyl Ester

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Melting Pt: 38 °C
MDL Number: MFCD01631193
CAS Number: 98977-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f06c837dd6dfa188d60cdc1b48b75872bd9939f5&vid=f06c837dd6dfa188d60cdc1b48b75872bd9939f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR