N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: Boc-Phe(4-F)-OH, N-Boc-4-fluoro-L-phenylalanine

B3304-1G B3304-5G
TCB3304-1GEA 104.28 CAD
TCB3304-1G TCB3304-5G
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine ≥98.0%
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine

CAS Number: 41153-30-4
MDL Number: MFCD00079672
Molecular Formula: C14H18FNO4
Molecular Weight: 283.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 85
Specific rotation [a]20/D: 19 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-4-fluoro-L-phenylalanine, Boc-Phe(4-F)-OH

Formula: C₁₄H₁₈FNO₄
Melting Pt: 85 °C
MDL Number: MFCD00079672
CAS Number: 41153-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7fee23fafd84dc930583844f81444678d7731c5&vid=b7fee23fafd84dc930583844f81444678d7731c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR