tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥98.0% (by GC)
Supplier: TCI America
B3273-1G
B3273-5G
TCB3273-1GEA
48.98
CAD
TCB3273-1G
TCB3273-5G
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥98.0% (by GC)
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate
CAS Number: 170491-63-1
MDL Number: MFCD01456556
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Other Product Names / Synonyms: N-Boc-DL-prolinol, 1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, 1-Boc-2-pyrrolidinemethanol
Formula:
C₁₀H₁₉NO₃ Melting Pt: 54 °C |
MDL Number:
MFCD01456556 CAS Number: 170491-63-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...