2-Bromo-6-methoxypyridine ≥97.0% (by GC)

Supplier: TCI America

B3260-25G B3260-5G
TCB3260-25GEA 191.18 CAD
TCB3260-25G TCB3260-5G
2-Bromo-6-methoxypyridine ≥97.0% (by GC)
2-Bromo-6-methoxypyridine

CAS Number: 40473-07-2
MDL Number: MFCD00088345
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 104
Specific Gravity (20/20): 1.55

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 91 °C
Density: 1.55 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00088345
CAS Number: 40473-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9111fe8d48130382ce3858b9c098367ce6d7534a&vid=9111fe8d48130382ce3858b9c098367ce6d7534a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR