8-Bromo-1-octene ≥98.0%

Supplier: TCI America

B3257-25G B3257-5G
TCB3257-25GEA 216.46 CAD
TCB3257-25G TCB3257-5G
8-Bromo-1-octene ≥98.0%
8-Bromo-1-octene

CAS Number: 2695-48-9
MDL Number: MFCD00000275
Molecular Formula: C8H15Br
Molecular Weight: 191.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 78
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: 7-Octenyl Bromide

Formula: C₈H₁₅Br
MW: 191.11
Boiling Pt: 92 °C
Density: 1.14 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00000275
CAS Number: 2695-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01ca4cd07091f7c904cccdb19e4298ab7744afed&vid=1b414afba65a5cbe0beeed1bfb05c68d27a98729&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR