1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Butyl-1,2-dimethyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide, 3-Butyl-1,2-dimethyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide

B3159-25G B3159-5G
TCB3159-25GEA 864.42 CAD
TCB3159-25G TCB3159-5G
1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)
1-Butyl-2,3-dimethylimidazolium bis(trifluoromethylsulphonyl)imide

CAS Number: 350493-08-2
Molecular Formula: C11H17F6N3O4S2
Molecular Weight: 433.38
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.43

Formula: C₁₁H₁₇F₆N₃O₄S₂
MW: 433.4 g/mol
Density: 1.43 (20 °C)
MDL Number: MFCD10566879
CAS Number: 350493-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=837c92a47e28f02d3c2612156c43e3fc90602ce6&vid=837c92a47e28f02d3c2612156c43e3fc90602ce6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR