1-Bromo-2-chloro-3-fluorobenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Chloro-3-fluorobromobenzene
B3119-25G
B3119-5G
TCB3119-25GEA
243.32
CAD
TCB3119-25G
TCB3119-5G
1-Bromo-2-chloro-3-fluorobenzene ≥98.0% (by GC)
1-Bromo-2-chloro-3-fluorobenzene
CAS Number: 883499-24-9
MDL Number: MFCD04038306
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 29
Formula:
C₆H₃BrClF MW: 209.44 g/mol |
MDL Number:
MFCD04038306 CAS Number: 883499-24-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...