(S)-(+)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)

Supplier: TCI America

B3055-1G B3055-5G
TCB3055-1GEA 50.56 CAD
TCB3055-1G TCB3055-5G
(S)-(+)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)
(S)-(+)-1-Boc-3-hydroxypyrrolidine

CAS Number: 101469-92-5
MDL Number: MFCD01317839
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63
Specific rotation [a]20/D: 27 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-1-Boc-3-pyrrolidinol, (S)-1-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine, (S)-1-Boc-3-hydroxypyrrolidine

Formula: C₉H₁₇NO₃
Melting Pt: 63 °C
MDL Number: MFCD01317839
CAS Number: 101469-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fbe974a297f4a93d45db0070be7570d9038dc75&vid=0fbe974a297f4a93d45db0070be7570d9038dc75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR