3-(Bromoacetyl)pyridine ydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B3006-25G
B3006-5G
TCB3006-25GEA
425.02
CAD
TCB3006-25G
TCB3006-5G
3-(Bromoacetyl)pyridine ydrobromide ≥98.0% (by HPLC, titration analysis)
3-(Bromoacetyl)pyridine ydrobromide
CAS Number: 17694-68-7
MDL Number: MFCD00052182
Molecular Formula: C7H6BrNO
Molecular Weight: 280.95
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 195
Storage Temperature: 0-10°C
Formula:
C₇H₆BrNO Melting Pt: 195 °C |
MDL Number:
MFCD00052182 CAS Number: 17694-68-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...