5-Bromo-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B2974-5G B2974-1G
TCB2974-5GEA 67.78 CAD
TCB2974-5G TCB2974-1G
5-Bromo-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
5-Bromo-2-fluorobenzoic acid

CAS Number: 146328-85-0
MDL Number: MFCD00143423
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 150

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00143423
CAS Number: 146328-85-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f621216b8f09b522ab72509299d7d0ec20c78d&vid=e9f621216b8f09b522ab72509299d7d0ec20c78d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR