2-Methylpropane-2-sulfinamide ≥97.0% (by GC)
Supplier: TCI America
B2926-1G
B2926-5G
TCB2926-1GEA
30.02
CAD
TCB2926-1G
TCB2926-5G
2-Methylpropane-2-sulfinamide ≥97.0% (by GC)
2-Methylpropane-2-sulfinamide
CAS Number: 146374-27-8
MDL Number: MFCD01863616
Molecular Formula: C4H11NOS
Molecular Weight: 121.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 107
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfinic Acid Derivatives
Other Product Names / Synonyms: 2-Methyl-2-propanesulfinamide
Formula:
C₄H₁₁NOS Melting Pt: 107 °C |
MDL Number:
MFCD01863616 CAS Number: 146374-27-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...