5-Bromo-2-chlorophenol ≥98.0% (by GC)

Supplier: TCI America

B2919-1G B2919-5G
TCB2919-1GEA 107.44 CAD
TCB2919-1G TCB2919-5G
5-Bromo-2-chlorophenol ≥98.0% (by GC)
5-Bromo-2-chlorophenol

CAS Number: 183802-98-4
MDL Number: MFCD00672940
Molecular Formula: C6H4BrClO
Molecular Weight: 207.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 234
Melting point (°C): 58

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Boiling Pt: 234 °C
Melting Pt: 58 °C
MDL Number: MFCD00672940
CAS Number: 183802-98-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d499ea2e93f4f21019628711d8bd0d055cc05df0&vid=d499ea2e93f4f21019628711d8bd0d055cc05df0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR