2-Bromo-α,α,α,5-tetrafluorotoluene ≥97.0% (by GC)

Supplier: TCI America

B2800-25G B2800-5G
TCB2800-25GEA 160.37 CAD
TCB2800-25G TCB2800-5G
2-Bromo-α,α,α,5-tetrafluorotoluene ≥97.0% (by GC)
2-Bromo-α,α,α,5-tetrafluorotoluene

CAS Number: 40161-55-5
MDL Number: MFCD00040937
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 64
Specific Gravity (20/20): 1.75


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 1-Bromo-4-fluoro-2-(trifluoromethyl)benzene

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 143 °C
Density: 1.75 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00040937
CAS Number: 40161-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50e95359141e693deba5879cfaf679c8a1eb145c&vid=50e95359141e693deba5879cfaf679c8a1eb145c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR