5-Bromo-2-thiophenesulfonamide ≥97.0%

Supplier: TCI America

Synonyms: 5-Bromothiophene-2-sulphonamide, 5-Bromothiophene-2-sulfonamide

B2793-25G B2793-5G
TCB2793-25GEA 186.44 CAD
TCB2793-25G TCB2793-5G
5-Bromo-2-thiophenesulfonamide ≥97.0%
5-Bromo-2-thiophenesulfonamide

CAS Number: 53595-65-6
MDL Number: MFCD00067990
Molecular Formula: C4H4BrNO2S2
Molecular Weight: 242.11
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 141

Formula: C₄H₄BrNO₂S₂
MW: 242.11 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00067990
CAS Number: 53595-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a3e2d12b478a98354c283aa40f01e43fe2fa253&vid=8a3e2d12b478a98354c283aa40f01e43fe2fa253&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR