1-Boc-isonipecotic acid ethyl ester ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate

B2740-25G B2740-5G
TCB2740-25GEA 178.54 CAD
TCB2740-25G TCB2740-5G
1-Boc-isonipecotic acid ethyl ester ≥97.0% (by GC)
1-Boc-isonipecotic acid ethyl ester

CAS Number: 142851-03-4
MDL Number: MFCD01763998
Molecular Formula: C13H23NO4
Molecular Weight: 257.33
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Boiling Pt: 140 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01763998
CAS Number: 142851-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8cde4eb822300a0ebcfb9ac0fd304f9fae2093f&vid=c8cde4eb822300a0ebcfb9ac0fd304f9fae2093f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR