Bromotrispyrrolidinophosphonium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B2680-25G B2680-5G
TCB2680-25GEA 383.94 CAD
TCB2680-25G TCB2680-5G
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate

CAS Number: 132705-51-2
MDL Number: MFCD00077412
Molecular Formula: C12H24BrF6N3P2
Molecular Weight: 466.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 139
Storage Temperature: <0°C

Formula: C₁₂H₂₄BrF₆N₃P₂
Melting Pt: 139 °C
MDL Number: MFCD00077412
CAS Number: 132705-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7d145f123eeafe2f0891dc4f9cc015c18db81d4&vid=f7d145f123eeafe2f0891dc4f9cc015c18db81d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR