2,2'-Bipyrimidyl ≥95.0% (by GC)
Supplier: TCI America
B2496-1G
B2496-200MG
TCB2496-1GEA
373.01
CAD
TCB2496-1G
TCB2496-200MG
2,2'-Bipyrimidyl ≥95.0% (by GC)
2,2'-Bipyrimidyl
CAS Number: 34671-83-5
MDL Number: MFCD00014600
Molecular Formula: C8H6N4
Molecular Weight: 158.16
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 113
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 2,2'-Bipyrimidine, 2,2'-Dipyrimidyl
Formula:
C₈H₆N₄ Melting Pt: 113 °C |
MDL Number:
MFCD00014600 CAS Number: 34671-83-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...