2-Biphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

B2488-25G B2488-5G
TCB2488-25GEA 443.98 CAD
TCB2488-25G TCB2488-5G
2-Biphenylboronic acid (contains varying amounts of Anhydride)
2-Biphenylboronic acid

CAS Number: 4688-76-0
MDL Number: MFCD00136929
Molecular Formula: C12H11BO2
Molecular Weight: 198.03
Form: Crystal
Color: White
Melting point (°C): 129
Lambda max.: 250 nm (CH2Cl2)

Formula: C₁₂H₁₁BO₂
Melting Pt: 129 °C
MDL Number: MFCD00136929
CAS Number: 4688-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f1976d8d5ae6b9b2de49f6c6648ba550c89cfb7&vid=3f1976d8d5ae6b9b2de49f6c6648ba550c89cfb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR