1-Butyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-Butyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide, 1-Butyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide, 1-Butyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide, 1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide
CAS Number: 174899-83-3
MDL Number: MFCD05664714
Molecular Formula: C10H15F6N3O4S2
Molecular Weight: 419.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Melting point (°C): -2
Specific Gravity (20/20): 1.44
Formula:
C₁₀H₁₅F₆N₃O₄S₂ MW: 419.37 g/mol Melting Pt: –2 °C Density: 1.44 (20 °C) |
MDL Number:
MFCD05664714 CAS Number: 174899-83-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...