1-Benzyloxy-3-bromobenzene ≥98.0%

Supplier: TCI America

Synonyms: 3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene

B2307-25G B2307-5G
TCB2307-25GEA 213.3 CAD
TCB2307-25G TCB2307-5G
1-Benzyloxy-3-bromobenzene ≥98.0%
1-Benzyloxy-3-bromobenzene

CAS Number: 53087-13-1
MDL Number: MFCD00155065
Molecular Formula: C13H11BrO
Molecular Weight: 263.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62


Other Product Names / Synonyms: Benzyl 3-Bromophenyl Ether

Formula: C₁₃H₁₁BrO
Melting Pt: 62 °C
MDL Number: MFCD00155065
CAS Number: 53087-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62e08bebf464f04cb789b01f454bea3951d73bb1&vid=62e08bebf464f04cb789b01f454bea3951d73bb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR