1-Benzyl-4-hydroxypiperidine ≥98.0%

Supplier: TCI America

Synonyms: 1-Benzylpiperidin-4-ol

B2252-25G B2252-5G
TCB2252-25GEA 161.16 CAD
TCB2252-25G TCB2252-5G
1-Benzyl-4-hydroxypiperidine ≥98.0%
1-Benzyl-4-hydroxypiperidine

CAS Number: 4727-72-4
MDL Number: MFCD00006503
Molecular Formula: C12H17NO
Molecular Weight: 191.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: 1-Benzyl-4-piperidinol

Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
MDL Number: MFCD00006503
CAS Number: 4727-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfcdfa7a2b9705817e033c4f8b65380683bfc109&vid=dfcdfa7a2b9705817e033c4f8b65380683bfc109&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR