[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride 97,0

Supplier: TCI America

Synonyms: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), [1,1'-Bis(diphenylphosphino)ferrocene]dichloronickel(II), [1,1’-Bis(diphenylphosphino)ferrocene]dichloronickel(II)

B2226-1G B2226-5G
TCB2226-1GEA 121.66 CAD
TCB2226-1G TCB2226-5G
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride 97,0
[1,1'-Bis(diphenylphosphino)ferrocene]nickel (II) chloride

CAS Number: 67292-34-6
MDL Number: MFCD00270284
Molecular Formula: C34H28Cl2FeNiP2
Molecular Weight: 683.98
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 259


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: [1,1'-Bis(diphenylphosphino)ferrocene]dichloronickel(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)

Formula: C₃₄H₂₈Cl₂FeNiP₂
MW: 683.98 g/mol
Melting Pt: 259 °C
MDL Number: MFCD00270284
CAS Number: 67292-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5298d613ceac809bdc5bacd6c6f60bc77171d2bd&vid=5298d613ceac809bdc5bacd6c6f60bc77171d2bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR