9,10-Bis(3,5-dihydroxyphenyl)anthracene ≥96.0%

Supplier: TCI America

B2109-100MG B2109-1G
TCB2109-100MGEA 143.78 CAD
TCB2109-100MG TCB2109-1G
9,10-Bis(3,5-dihydroxyphenyl)anthracene ≥96.0%
9,10-Bis(3,5-dihydroxyphenyl)anthracene

CAS Number: 153715-08-3
MDL Number: MFCD03427350
Molecular Formula: C26H18O4
Molecular Weight: 394.43
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal

Formula: C₂₆H₁₈O₄
MDL Number: MFCD03427350
CAS Number: 153715-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e618ffa16a62922d5f1bb8d26d0405d1c81aa3e&vid=5e618ffa16a62922d5f1bb8d26d0405d1c81aa3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR