4-Bromo-2,6-dichloroaniline ≥98.0%

Supplier: TCI America

B1978-25G B1978-5G
TCB1978-25GEA 259.12 CAD
TCB1978-25G TCB1978-5G
4-Bromo-2,6-dichloroaniline ≥98.0%
4-Bromo-2,6-dichloroaniline

CAS Number: 697-88-1
MDL Number: MFCD00025162
Molecular Formula: C6H4BrCl2N
Molecular Weight: 240.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83

Formula: C₆H₄BrCl₂N
Melting Pt: 83 °C
MDL Number: MFCD00025162
CAS Number: 697-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c3076c4fb29f323258fc265576cbaef65c7e4c8&vid=8929130f8181fbe8fc71633f75bd9fc9a0850ff8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR