4-Bromo-1-butanol ≥80.0% (contains varying amounts of Tetrahydrofuran)

Supplier: TCI America

Synonyms: 1-Bromo-4-hydroxybutane

B1885-25G B1885-5G
TCB1885-25GEA 560.9 CAD
TCB1885-25G TCB1885-5G
4-Bromo-1-butanol ≥80.0% (contains varying amounts of Tetrahydrofuran)
4-Bromo-1-butanol

CAS Number: 33036-62-3
MDL Number: MFCD00015387
Molecular Formula: C4H9BrO
Molecular Weight: 153.02
Purity/Analysis Method: >80.0% (NMR)
Form: Clear Liquid
Storage Temperature: <0°C

Formula: BrH₂C(CH₂)₂CH₂OH
MDL Number: MFCD00015387
CAS Number: 33036-62-3
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c78a57976555fce4f4f81ef9c9598ace5e44e0d4&vid=c78a57976555fce4f4f81ef9c9598ace5e44e0d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR